Skip to Content
MilliporeSigma
All Photos(2)

Documents

483370

Sigma-Aldrich

2,6-Bis(2-pyridyl)-4(1H)-pyridone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11N3O
CAS Number:
Molecular Weight:
249.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

168-170 °C (lit.)

SMILES string

O=C1C=C(NC(=C1)c2ccccn2)c3ccccn3

InChI

1S/C15H11N3O/c19-11-9-14(12-5-1-3-7-16-12)18-15(10-11)13-6-2-4-8-17-13/h1-10H,(H,18,19)

InChI key

HRORSVNZQWCZTD-UHFFFAOYSA-N

Application

2,6-Bis(2-pyridyl)-4(1H)-pyridone can be used a heterocyclic building block to prepare 4′-substituted terpyridines by reacting with ω-substituted primary alcohols or nucleosides via Mitsunobu reaction. It is also used to prepare cyclotriphosphazene ligands with pendant terpyridine moieties.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toward an iron (II) spin-crossover grafted phosphazene polymer
Davidson RJ, et al.
Inorganic Chemistry, 51(15), 8307-8316 (2012)
Synthesis of 4?-substituted 2, 2?: 6?, 2 ?-terpyridines via a Mitsunobu reaction
Hovinen J
Tetrahedron Letters, 45(29), 5707-5709 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service