Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47691

Sigma-Aldrich

Fmoc-α-Me-Ala-OH

≥97.0% (HPLC)

Synonym(s):

Fmoc-α-aminoisobutyric acid, Fmoc-α-methylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H19NO4
CAS Number:
Molecular Weight:
325.36
Beilstein:
5604328
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Quality Level

Assay

≥97.0% (HPLC)

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

182-188 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

CC(C)(NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C19H19NO4/c1-19(2,17(21)22)20-18(23)24-11-16-14-9-5-3-7-12(14)13-8-4-6-10-15(13)16/h3-10,16H,11H2,1-2H3,(H,20,23)(H,21,22)

InChI key

HOZZVEPRYYCBTO-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aleksandra Balliu et al.
Chembiochem : a European journal of chemical biology, 18(14), 1396-1407 (2017-04-23)
A 42-residue polypeptide conjugated to a small-molecule organic ligand capable of targeting the phosphorylated side chain of Ser15 was shown to bind glycogen phosphorylase a (GPa) with a K

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service