Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47634

Sigma-Aldrich

Fmoc-Met-OH

≥98.0% (HPLC), for peptide synthesis

Synonym(s):

Fmoc-L-methionine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21NO4S
CAS Number:
Molecular Weight:
371.45
Beilstein:
4300266
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

Fmoc-Met-OH, ≥98.0% (HPLC)

Assay

≥98.0% (HPLC)

optical activity

[α]20/D -29.5±1.5°, c = 1% in DMF

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

121-123 °C (lit.)

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

CSCC[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C20H21NO4S/c1-26-11-10-18(19(22)23)21-20(24)25-12-17-15-8-4-2-6-13(15)14-7-3-5-9-16(14)17/h2-9,17-18H,10-12H2,1H3,(H,21,24)(H,22,23)/t18-/m0/s1

InChI key

BUBGAUHBELNDEW-SFHVURJKSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dennis Kubiczek et al.
Macromolecular bioscience, 20(4), e2000005-e2000005 (2020-02-28)
The pathogenic yeast Candida auris has received increasing attention due to its ability to cause fatal infections, its resistance toward important fungicides, and its ability to persist on surfaces including medical devices in hospitals. To brace health care systems for
Carolyn C Woodroofe et al.
Chembiochem : a European journal of chemical biology, 21(4), 508-516 (2019-08-01)
The reversible oxidation of methionine residues in proteins has emerged as a biologically important post-translational modification. However, detection and quantitation of methionine sulfoxide in proteins is difficult. Our aim is to develop a method for specifically derivatizing methionine sulfoxide residues.
Marcus Pickhardt et al.
Current Alzheimer research, 14(7), 742-752 (2017-02-06)
Anti-aggregation drugs play an important role in therapeutic approaches for Alzheimer's disease. We have previously developed a number of compounds that are able to inhibit the pathological aggregation of Tau protein. One common obstacle to application is the limited penetration
Gizella Csire et al.
Journal of inorganic biochemistry, 170, 195-201 (2017-03-07)
The prion protein (PrP) is a membrane-anchored cell surface glycoprotein containing 231 amino acids. It has been associated with a group of neurodegenerative disorders. Copper(II) interaction with the Human Prion 103-112 fragment and its mutants has been studied with various
Gizella Csire et al.
Journal of inorganic biochemistry, 203, 110927-110927 (2019-12-07)
Interaction of copper(II) and nickel(II) ions with the Ac-PHAAAGTHSMKHM-NH2 tridecapeptide containing the His85, His96 and His111 binding sites of human prion protein has been studied by various techniques. pH-potentiometry, UV-Vis and circular dichroism spectroscopy were applied to study the stoichiometry

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service