Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47601

Sigma-Aldrich

Fmoc-Ser-OH

≥97.0% (sum of enantiomers, HPLC)

Synonym(s):

Fmoc-L-serine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17NO5
CAS Number:
Molecular Weight:
327.33
Beilstein:
4715791
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (sum of enantiomers, HPLC)

optical activity

[α]20/D −12.5±1°, c = 1% in DMF

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C18H17NO5/c20-9-16(17(21)22)19-18(23)24-10-15-13-7-3-1-5-11(13)12-6-2-4-8-14(12)15/h1-8,15-16,20H,9-10H2,(H,19,23)(H,21,22)/t16-/m0/s1

InChI key

JZTKZVJMSCONAK-INIZCTEOSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annabelle Biscans et al.
Nucleic acids research, 47(3), 1082-1096 (2018-12-14)
Small interfering RNA (siRNA)-based therapies are proving to be efficient for treating liver-associated disorders. However, extra-hepatic delivery remains challenging, limiting therapeutic siRNA utility. We synthesized a panel of fifteen lipid-conjugated siRNAs and systematically evaluated the impact of conjugate on siRNA
Frans J Walther et al.
PloS one, 5(1), e8672-e8672 (2010-01-20)
Surfactant protein B (SP-B; 79 residues) belongs to the saposin protein superfamily, and plays functional roles in lung surfactant. The disulfide cross-linked, N- and C-terminal domains of SP-B have been theoretically predicted to fold as charged, amphipathic helices, suggesting their
Jingwen Song et al.
Small (Weinheim an der Bergstrasse, Germany), 16(8), e1907309-e1907309 (2020-01-30)
There is a real need for new antibiotics against self-evolving bacteria. One option is to use biofriendly broad-spectrum and mechanically tunable antimicrobial hydrogels that can combat multidrug-resistant microbes. Whilst appealing, there are currently limited options. Herein, broad-spectrum antimicrobial biometallohydrogels based
Peiming Chen et al.
Journal of controlled release : official journal of the Controlled Release Society, 194, 341-349 (2014-09-16)
Macrophages are involved in a number of diseases, such as HIV infection/AIDS, tuberculosis, tumor development and atherosclerosis. Macrophages possess several cell surface receptors (e.g., the mannose receptor, MR) that may serve as drug delivery cellular portals for nanocarriers (NCs). In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service