Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

471526

Sigma-Aldrich

1-Dimethylamino-2-propanol

≥99%

Synonym(s):

N,N-Dimethylisopropanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2N(CH3)2
CAS Number:
Molecular Weight:
103.16
Beilstein:
1209244
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.6 (vs air)

Quality Level

vapor pressure

8 mmHg ( 20 °C)

Assay

≥99%

refractive index

n20/D 1.419 (lit.)

bp

121-127 °C (lit.)

density

0.837 g/mL at 25 °C (lit.)

SMILES string

CC(O)CN(C)C

InChI

1S/C5H13NO/c1-5(7)4-6(2)3/h5,7H,4H2,1-3H3

InChI key

NCXUNZWLEYGQAH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Dimethylamino-2-propanol (1DMA2P, DMAPH), a tertiary amine, is a dimethylamino-alcohol having high boiling point. It is a potent protector against mechlorethamine cytotoxicity and inhibitor of choline uptake. Kinetics of homogeneous reaction of carbon dioxide (CO2) with 1-dimethylamino-2-propanol in water has been investigated using stopped-flow technique.

Application

1-Dimethylamino-2-propanol solution may be used in the synthesis of the following:
  • novel unsymmetrical 2,3,9,10,16,17,23-heptakis(alkoxyl)-24-mono(dimethylaminoalkoxyl)phthalocyanine compounds
  • homoleptic nickel(II) aminoalkoxide
  • Adduct with cobalt(II) 2,4-pentanedionate (acac)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

78.8 °F - DIN 51755 Part 1

Flash Point(C)

26 °C - DIN 51755 Part 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soluble Ni II alkoxides based on dimethylaminoisopropoxide ligands: molecular structure of [Li (Pr i OH) Ni (η 2-OR)2 Cl]2 and of cis-NiCl2 (ROH)2 (R= CHMeCH2NMe2).
Hubert-Pfalzgraf LG, et al.
Polyhedron, 16(24), 4197-4203 (1997)
Kinetics of carbon dioxide (CO 2) with ethylenediamine, 3-amino-1-propanol in methanol and ethanol, and with 1-dimethylamino-2-propanol and 3-dimethylamino-1-propanol in water using stopped-flow technique
Kadiwala S, et al.
Chemical Engineering Journal, 179, 262-271 (2012)
Zhaopin Bai et al.
Inorganic chemistry, 49(19), 9005-9011 (2010-09-04)
A new pathway by means of transetherification has been developed to synthesize novel unsymmetrical 2,3,9,10,16,17,23-heptakis(alkoxyl)-24-mono(dimethylaminoalkoxyl)phthalocyanine compounds. Cyclic tetramerization of 4,5-di(alkoxyl)phthalonitrile in refluxing dimethylamino-alcohol with high boiling point such as dimethylaminoethanol (DMAE) and 1-dimethylamino-2-propanol in the presence of lithium and pyrazino[2,3-f][1,10]phenanthroline-2,3-dicarbonitrile
S A Naujokaitis et al.
Journal of pharmaceutical sciences, 73(1), 34-39 (1984-01-01)
The structure-activity relationships of 2-dimethylaminoethanol and its analogues as protectors against mechlorethamine cytotoxicity and as inhibitors of choline uptake were evaluated. Of a series of inhibitors and protectors, 2-dimethylaminoethanol was the most potent inhibitor of choline uptake and the most
Synthesis and molecular structures of cobalt (II) ?-diketonate complexes as new MOCVD precursors for cobalt oxide films.
Pasko S, et al.
Polyhedron, 23(5), 735-741 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service