Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

466468

Sigma-Aldrich

N-(tert-Butoxycarbonyl)-L-valine methyl ester

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH[NHCO2C(CH3)3]CO2CH3
CAS Number:
Molecular Weight:
231.29
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]20/D −22°, c = 1 in methanol

reaction suitability

reaction type: solution phase peptide synthesis

refractive index

n20/D 1.44 (lit.)

bp

194 °C (lit.)

density

1.004 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

SMILES string

COC(=O)[C@@H](NC(=O)OC(C)(C)C)C(C)C

InChI

1S/C11H21NO4/c1-7(2)8(9(13)15-6)12-10(14)16-11(3,4)5/h7-8H,1-6H3,(H,12,14)/t8-/m0/s1

InChI key

XCJLIYKAMLUDGN-QMMMGPOBSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service