Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

464600

Sigma-Aldrich

Zirconium(IV) acetate hydroxide

Synonym(s):

Dihydroxy-bis(methoxycarbonyloxy)zirconium, Hydroxy-tris(methoxycarbonyloxy)zirconium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO2)xZr(OH)y, x+y ~4
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

composition

Zr, 40.4-43.3% gravimetric

reaction suitability

core: zirconium

SMILES string

COC(=O)O[Zr](O)(O)O.COC(=O)O[Zr](O)(O)OC(=O)OC.COC(=O)O[Zr](O)(OC(=O)OC)OC(=O)OC

InChI

1S/6C2H4O3.6H2O.3Zr/c6*1-5-2(3)4;;;;;;;;;/h6*1H3,(H,3,4);6*1H2;;;/q;;;;;;;;;;;;3*+4/p-12

InChI key

RAVMIOKPFXMREI-UHFFFAOYSA-B

Application

Zirconium(IV) acetate hydroxide can be used as a precursor to synthesize:
  • Monoclinic zirconium dioxide nanoparticles via thermal treatment method in the presence of polyvinylpyrrolidone as a capping agent.
  • Zirconia doped lithium cobalt oxide (Zr-LiCoO2) by treating with lithium acetate and cobalt acetate via the sol-gel method.
  • Yttrium stabilized zirconia nanoparticles via co-precipitation method using yttrium acetate tetrahydrate and oxalic acid dehydrate.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of yttrium stabilized zirconia nanoparticles
Maridurai T, et al.
Materials Research, 19, 812-816 (2016)
Structural and optical properties of zirconia nanoparticles by thermal treatment synthesis
Keiteb Aysar S, et al.
Journal of Nanomaterials, 2016 (2016)
Electrochemical properties of LiMxCo1- xO2 [M= Mg, Zr] prepared by sol-gel process
Kim Hyung-Sun, et al.
Journal of Power Sources, 138(1-2), 232-239 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service