Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

46131

Sigma-Aldrich

Methyl 2-ethylacetoacetate

≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH(C2H5)COOCH3
CAS Number:
Molecular Weight:
144.17
Beilstein:
1754068
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
≥97.0% (GC)

Quality Level

Assay

≥97.0% (GC)

refractive index

n20/D 1.423

density

1.010 g/mL at 20 °C (lit.)

functional group

ester
ketone

SMILES string

CCC(C(C)=O)C(=O)OC

InChI

1S/C7H12O3/c1-4-6(5(2)8)7(9)10-3/h6H,4H2,1-3H3

InChI key

YXLVLOWNJCOOAU-UHFFFAOYSA-N

General description

Methyl 2-ethylacetoacetate participates in the preparation of 5,6-disubstituted-1,3-dioxine-4-one.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nils Pemberton et al.
Organic letters, 8(5), 935-938 (2006-02-24)
Polycyclic ring-fused 2-pyridones (5a-e and 9a-e) have been prepared via a microwave-assisted acyl-ketene imine cyclocondensation. Starting from 3,4-dihydroisoquinolines (4a-b) or 3,4-dihydroharman (8), fused 2-pyridones could be prepared in a one-step procedure. By using either Meldrum's acid derivatives (1a-d) or 1,3-dioxine-4-ones

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service