Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

459232

Sigma-Aldrich

L-Valinamide hydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH(NH2)CONH2 · HCl
CAS Number:
Molecular Weight:
152.62
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

optical activity

[α]25/D +25.8°, c = 1 in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

266-270 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cl.CC(C)[C@H](N)C(N)=O

InChI

1S/C5H12N2O.ClH/c1-3(2)4(6)5(7)8;/h3-4H,6H2,1-2H3,(H2,7,8);1H/t4-;/m0./s1

InChI key

XFCNYSGKNAWXFL-WCCKRBBISA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miki Hasegawa et al.
ChemPlusChem, 85(2), 294-300 (2020-01-23)
Ligands based on 2,2'-bipyridine and valinamide moieties induce circularly polarized luminescence in their europium complexes. Both the R and S enantiomers of the complexes were successfully obtained. Single-crystal X-ray analysis of the racemic crystal confirmed that the ligand is coordinated
Griet Van Zeebroeck et al.
Nature chemical biology, 5(1), 45-52 (2008-12-09)
Transporter-related nutrient sensors, called transceptors, mediate nutrient activation of signaling pathways through the plasma membrane. The mechanism of action of transporting and nontransporting transceptors is unknown. We have screened 319 amino acid analogs to identify compounds that act on Gap1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service