Skip to Content
MilliporeSigma
L-Valinamide hydrochloride 97%

L-Valinamide hydrochloride

L-Valinamide hydrochloride 97%
Linear Formula:
(CH3)2CHCH(NH2)CONH2 · HCl
CAS Number:
Molecular Weight:
152.62

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing