Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

456926

Sigma-Aldrich

1-Z-Piperazine

98%

Synonym(s):

1-(Benzyloxycarbonyl)piperazine, 1-Cbz-Piperazine, Benzyl piperazine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16N2O2
CAS Number:
Molecular Weight:
220.27
Beilstein:
179500
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.546 (lit.)

bp

158-161 °C/1.4 mmHg (lit.)

density

1.142 g/mL at 25 °C (lit.)

functional group

phenyl

SMILES string

O=C(OCc1ccccc1)N2CCNCC2

InChI

1S/C12H16N2O2/c15-12(14-8-6-13-7-9-14)16-10-11-4-2-1-3-5-11/h1-5,13H,6-10H2

InChI key

CTOUWUYDDUSBQE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dai Nozawa et al.
Bioorganic & medicinal chemistry, 15(6), 2375-2385 (2007-02-03)
In the present study, we found that a novel piperazine compound, 11a, showed a moderate affinity (IC(50)=333nM) for the MC4 receptor. We developed the new type of piperazine compounds and found that mono-piperazine 11b exhibited a high-affinity (IC(50)=40.3nM) for the
Synthesis of Macrocyclic, Triazine-Based Receptor Molecules.
Lowik DWPM and Lowe CR.
European Journal of Organic Chemistry, 2001(15), 2825-2839 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service