Skip to Content
MilliporeSigma
All Photos(2)

Documents

456195

Sigma-Aldrich

1-Bromo-2-(2-methoxyethoxy)ethane

90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(OCH2CH2)2Br
CAS Number:
Molecular Weight:
183.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

refractive index

n20/D 1.455 (lit.)

bp

156 °C (lit.)
37-40 °C/0.01 mmHg (lit.)

density

1.3600 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

COCCOCCBr

InChI

1S/C5H11BrO2/c1-7-4-5-8-3-2-6/h2-5H2,1H3

InChI key

HUXJXNSHCKHFIL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.9 °F - closed cup

Flash Point(C)

68.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular engineering of photoremovable protecting groups for two-photon uncaging.
Sylvestre Gug et al.
Angewandte Chemie (International ed. in English), 47(49), 9525-9529 (2008-10-31)
E Córdova et al.
Analytical chemistry, 69(7), 1370-1379 (1997-04-01)
The adsorption of proteins with net positive charges (pI > pH) on the walls of fused-silica capillaries is a common problem in the analysis of proteins by capillary electrophoresis. This paper explores the use of polycationic polymers as noncovalent coatings
Effect of charge density on energy-transfer properties of cationic conjugated polymers.
Pu K-Y, et al.
Advances in Functional Materials, 18(8), 1321-1328 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service