Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

454850

Sigma-Aldrich

2-Methyl-4-nitropyridine N-oxide

97%

Synonym(s):

4-Nitro-2-picoline N-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N2O3
CAS Number:
Molecular Weight:
154.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

155-159 °C (lit.)

functional group

nitro

SMILES string

Cc1cc(cc[n+]1[O-])[N+]([O-])=O

InChI

1S/C6H6N2O3/c1-5-4-6(8(10)11)2-3-7(5)9/h2-4H,1H3

InChI key

FTTIAVRPJGCXAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Max Peukert et al.
Acta crystallographica. Section E, Structure reports online, 70(Pt 4), o426-o427 (2014-05-16)
An improved crystal structure of the title compound, C6H6N2O3, is reported. The structure, previously solved [Li et al. (1987 ▶). Jiegou Huaxue (Chin. J. Struct. Chem.), 6, 20-24] in the ortho-rhom-bic space group Pca21 and refined to R = 0.067
Aniela Puszko et al.
Journal of inorganic biochemistry, 104(2), 153-160 (2009-11-26)
Three new complexes, products of the interaction of Cu(NO(3))(2) and methyl-substituted 4-nitropyridine N-oxides were synthesized and characterized by elemental analysis, magnetic, spectroscopic (IR, FIR and EPR), thermal and X-ray methods. The complexes (magnetic moments 1.70-1.81 BM at 300K) of general
Amination of alkyl substituted halogenopyridines with potassium amide in liquid ammonia.
Van der Does L and Den Hertog HJ.
Rec. Trav. Chim., 91(12), 1403-1413 (1972)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service