Skip to Content
MilliporeSigma
All Photos(1)

Documents

44230

Sigma-Aldrich

Trichlorododecylsilane

≥95.0% (GC)

Synonym(s):

Dodecyltrichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11SiCl3
CAS Number:
Molecular Weight:
303.77
Beilstein:
1758004
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (GC)

form

liquid

refractive index

n20/D 1.454 (lit.)
n20/D 1.454

bp

293-294 °C (lit.)

density

1.02 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCC[Si](Cl)(Cl)Cl

InChI

1S/C12H25Cl3Si/c1-2-3-4-5-6-7-8-9-10-11-12-16(13,14)15/h2-12H2,1H3

InChI key

BNCXNUWGWUZTCN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adem Yildirim et al.
ACS applied materials & interfaces, 10(43), 36786-36795 (2018-10-20)
While thermal ablation of various solid tumors has been demonstrated using high intensity focused ultrasound (HIFU), the therapeutic outcomes of this technique are still unsatisfactory because of common recurrence of thermally ablated cancers and treatment side effects due to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service