Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M85301

Sigma-Aldrich

Methyltrichlorosilane

99%

Synonym(s):

MTS, Trichloro(methyl)silane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SiCl3
CAS Number:
Molecular Weight:
149.48
Beilstein:
1361381
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.2 (vs air)

Quality Level

vapor pressure

150 mmHg ( 25 °C)

Assay

99%

form

liquid

autoignition temp.

>760 °F

expl. lim.

11.9 %

refractive index

n20/D 1.411 (lit.)

bp

66 °C (lit.)

density

1.273 g/mL at 25 °C (lit.)

SMILES string

C[Si](Cl)(Cl)Cl

InChI

1S/CH3Cl3Si/c1-5(2,3)4/h1H3

InChI key

JLUFWMXJHAVVNN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyltrichlorosilane (MTS) can be used:
  • As a reagent for the surface-functionalization of fabrics and mesoporous materials to increase their hydrophobicity.
  • As a source of silicon to synthesize silica doped carbon nanotubes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

46.4 °F - closed cup

Flash Point(C)

8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ville Liimatainen et al.
Nature communications, 8(1), 1798-1798 (2017-11-28)
Droplets slip and bounce on superhydrophobic surfaces, enabling remarkable functions in biology and technology. These surfaces often contain microscopic irregularities in surface texture and chemical composition, which may affect or even govern macroscopic wetting phenomena. However, effective ways to quantify
Mesoporous SiO2-TiO2 epoxidation catalysts: Tuning surface polarity to improve performance in the presence of water.
Smeets V, et al.
Molecular Catalysis, 452, 123-128 (2018)
Lishen Zhang et al.
ACS applied materials & interfaces, 9(45), 39728-39735 (2017-09-22)
Off-the-shelf laboratory filter paper of different pore sizes and thicknesses can be modified with fluorine-free organosilanes to be superhydrophobic, patternable, and ready for quantitative assay applications. In particular, we have demonstrated that the cellulose filter paper treated with a binary
Jianhua Wang et al.
PloS one, 11(3), e0151439-e0151439 (2016-03-18)
In this paper, we present a facile and cost-effective method to obtain superhydrophobic filter paper and demonstrate its application for efficient water/oil separation. By coupling structurally distinct organosilane precursors (e.g., octadecyltrichlorosilane and methyltrichlorosilane) to paper fibers under controlled reaction conditions
A novel strategy for fabricating robust superhydrophobic fabrics by environmentally-friendly enzyme etching.
Cheng Y, et al.
Chemical Engineering Journal, 355, 290-298 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service