Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

441627

Sigma-Aldrich

3-Bromophenylboronic acid

≥95%

Synonym(s):

3-Bromobenzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4B(OH)2
CAS Number:
Molecular Weight:
200.83
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

powder or crystals

mp

164-168 °C (lit.)

functional group

bromo

SMILES string

OB(O)c1cccc(Br)c1

InChI

1S/C6H6BBrO2/c8-6-3-1-2-5(4-6)7(9)10/h1-4,9-10H

InChI key

AFSSVCNPDKKSRR-UHFFFAOYSA-N

Application

Reactant involved in a variety of organic reactions including:
  • Oxidative cross coupling
  • Gold salt catalyzed homocoupling
  • 1,4-Addition reactions with α,β-unsaturated ketones
  • Enantioselective addition reactions
  • Suzuki-Miyaura coupling for synthesis of anthranilamide-protected arylboronic acids
  • C-H Functionalization of quinones

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G K Surya Prakash et al.
Organic letters, 6(13), 2205-2207 (2004-06-18)
[reaction: see text] A mixture of nitrate salt and chlorotrimethylsilane is found to be an efficient regioselective nitrating agent for the ipso-nitration of arylboronic acids to produce the corresponding nitroarenes in moderate to excellent yields. High selectivity, simplicity, and convenience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service