Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

900988

Sigma-Aldrich

3-Aminophenylboronic acid

Synonym(s):

3-Aminobenzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8BNO2
CAS Number:
Molecular Weight:
136.94
EC Number:
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22
Pricing and availability is not currently available.

form

powder or crystals

Quality Level

mp

225 °C

SMILES string

Nc1cccc(c1)B(O)O

InChI

1S/C6H8BNO2/c8-6-3-1-2-5(4-6)7(9)10/h1-4,9-10H,8H2

InChI key

JMZFEHDNIAQMNB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Aminophenylboronic acid (3-APBA) can undergo three-component condensation with 3,5-disubstituted salicylaldehyde derivatives and aliphatic alcohols to afford calix-shaped boron complexes.[1] Due to its ability to selectively bind with diol-compounds, 3-APBA can be employed in developing sensors for detecting saccharides and glycosylated biomolecules by immobilizing on a metal substrate via a self-assembled monolayer.[2] This product was previously listed as CDS003827.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rodtichoti Wannapob et al.
Biosensors & bioelectronics, 26(2), 357-364 (2010-08-31)
Boronic acid that can reversibly bind to diols was used to detect bacteria through its affinity binding reaction with diol-groups on bacterial cell walls. 3-aminophenylboronic acid (3-APBA) was immobilized on a gold electrode via a self-assembled monolayer. The change in
3-Aminophenylboronic acid as building block for the construction of calix- and cage-shaped boron complexes.
Barba V, et al.
Journal of Organometallic Chemistry, 694, 2127-2133 (2009)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service