Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

440116

Sigma-Aldrich

Dichloro(methyl)phenylsilane

97%

Synonym(s):

Methyl-phenyl-dichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5Si(CH3)Cl2
CAS Number:
Molecular Weight:
191.13
Beilstein:
970975
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.519 (lit.)

bp

205 °C (lit.)

density

1.176 g/mL at 25 °C (lit.)

SMILES string

C[Si](Cl)(Cl)c1ccccc1

InChI

1S/C7H8Cl2Si/c1-10(8,9)7-5-3-2-4-6-7/h2-6H,1H3

InChI key

GNEPOXWQWFSSOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service