Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

435937

Sigma-Aldrich

Cerium(IV) fluoride

99%

Synonym(s):

Ceric fluoride, Cerium tetrafluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CeF4
CAS Number:
Molecular Weight:
216.11
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

powder

reaction suitability

core: cerium
reagent type: catalyst
reaction type: Fluorinations

SMILES string

F[Ce](F)(F)F

InChI

1S/Ce.4FH/h;4*1H/q+4;;;;/p-4

InChI key

SWURHZJFFJEBEE-UHFFFAOYSA-J

Application

Reactant for:
  • Synthesis of sodium-rare earth silicate oxide-fluoride apatites
  • C60 fluorination with rare earth metal tetrafluorides
  • Synthesis of cerium(IV) fluoride compounds

Reagent for:
  • Fluorination of fluorocyclobutene with high-valency metal fluoride

Catalyst for Hantzsch reaction

Used for stain etching to form microporous silicon
Employed in the fluorination of hydrocarbons.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zh. Org. Khim., 30, 1221-1221 (1994)

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service