434116
Tetraethylammonium hexafluorophosphate
99%
Synonym(s):
N,N,N-triethyl-ethanaminium hexafluorophosphate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C2H5)4N(PF6)
CAS Number:
Molecular Weight:
275.22
Beilstein:
3748868
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
mp
≥300 °C (lit.)
SMILES string
F[P-](F)(F)(F)(F)F.CC[N+](CC)(CC)CC
InChI
1S/C8H20N.F6P/c1-5-9(6-2,7-3)8-4;1-7(2,3,4,5)6/h5-8H2,1-4H3;/q+1;-1
InChI key
KLKUOIXSIDDDCN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Employed extensively in electrolytes for electrochemical synthesis. Recent examples include one-pot electrochemical generation of a porphyrin dimer and reduction of esters in aprotic solvents.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
The Journal of Physical Chemistry, 100, 10288-10288 (1996)
Chemical Communications (Cambridge, England), 2007-2007 (1996)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service