Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
(C2H5)4N(PF6)
CAS Number:
Molecular Weight:
275.22
UNSPSC Code:
26111700
NACRES:
NB.61
PubChem Substance ID:
EC Number:
207-056-7
Beilstein/REAXYS Number:
3748868
MDL number:
Quality Level
assay
≥99.0% (T), ≥99.0%
form
crystals
mp
≥300 °C (lit.)
solubility
acetonitrile: 0.1 g/mL, clear, colorless
SMILES string
F[P-](F)(F)(F)(F)F.CC[N+](CC)(CC)CC
InChI
1S/C8H20N.F6P/c1-5-9(6-2,7-3)8-4;1-7(2,3,4,5)6/h5-8H2,1-4H3;/q+1;-1
InChI key
KLKUOIXSIDDDCN-UHFFFAOYSA-N
General description
Visit our Sensor Applications portal to learn more.
Application
Supporting electrolyte
Still not finding the right product?
Explore all of our products under Tetraethylammonium hexafluorophosphate
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 86625-25G | 04061833041642 |
