Skip to Content
MilliporeSigma
All Photos(2)

Documents

432393

Sigma-Aldrich

Polystyrene-block-polyisoprene-block-polystyrene

styrene 14 wt. %

Synonym(s):

Polystyrene-block-polyisoprene, multi-arm

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H5)]X[CH2CH=C(CH3)CH2]Y[CH2CH(C6H5)]z
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

beads

melt index

11 g/10 min (200°C/5.0kg)

contains

phenolic as antioxidant

composition

styrene, 14 wt. %

hardness

37 (Shore A)

viscosity

14.5 poise, 25 wt. % in toluene(25 °C, Brookfield)(lit.)

solubility

solvents with solubility parameters between 7.7 and 9.4: soluble

density

0.92 g/mL at 25 °C

elongation

ASTM D 412 - 1,300% (ultimate)

SMILES string

CC(=C)C=C.C=Cc1ccccc1

InChI

1S/C8H8.C5H8/c1-2-8-6-4-3-5-7-8;1-4-5(2)3/h2-7H,1H2;4H,1-2H2,3H3

InChI key

ROGIWVXWXZRRMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Component of hot melt adhesives, sealants, asphalt and oil gels. Modifier for polymers, thermosets and general rubber compounding. Direct molded or extruded auto parts, sporting goods, footware and film.

Features and Benefits

High strength and low viscosity for easy melt and solution processing. High temperature degradation results in cross-linking. Flexible to -65°C. Excellent electrical insulator, resists acids and alkali.

Physical form

Thermoplastic elastomer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service