Skip to Content
MilliporeSigma
Polystyrene-block-polyisoprene-block-polystyrene styrene 17 wt. %

Polystyrene-block-polyisoprene-block-polystyrene

Polystyrene-block-polyisoprene-block-polystyrene styrene 17 wt. %
Synonym(s):
Polystyrene-block-polyisoprene, multi-arm
Linear Formula:
[CH2CH(C6H5)]X[CH2CH=C(CH3)CH2]Y[CH2CH(C6H5)]z
CAS Number:

Products

Brand
Physical Form
Chemical Composition
Polymer Composition
Polymer Type