Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

415111

Sigma-Aldrich

Magnesium nitride

cubic phase, −325 mesh, ≥99.5% trace metals basis

Synonym(s):

Trimagnesium dinitride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg3N2
CAS Number:
Molecular Weight:
100.93
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.5% trace metals basis

form

powder

particle size

−325 mesh

density

2.71 g/mL at 25 °C (lit.)

SMILES string

[Mg]1[Mg]N=N[Mg]1

InChI

1S/3Mg.N2/c;;;1-2/q;2*+1;-2

InChI key

VZPQZBKNAUHPOS-UHFFFAOYSA-N

Related Categories

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3 - Water-react 1

Target Organs

Respiratory system

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Olivia Wenzel et al.
Microscopy and microanalysis : the official journal of Microscopy Society of America, Microbeam Analysis Society, Microscopical Society of Canada, 26(1), 102-111 (2020-01-11)
Nanoporous, high-purity magnesium nitride (Mg3N2) was synthesized with a liquid ammonia-based process, for potential applications in optoelectronics, gas separation and catalysis, since these applications require high material purity and crystallinity, which has seldom been demonstrated in the past. One way

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service