Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

404926

Sigma-Aldrich

Vanadium(V) oxytriisopropoxide

Synonym(s):

Triisopropoxyvanadium(V) oxide, VTIP, Vanadium(V) trisisopropoxide oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OV(OCH(CH3)2)3
CAS Number:
Molecular Weight:
244.20
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

liquid

composition

V, 20.3-21.4% titration (by potassium permanganate)

reaction suitability

core: vanadium
reagent type: catalyst

refractive index

n20/D 1.479 (lit.)

bp

80-82 °C/2 mmHg (lit.)

density

1.035 g/mL at 25 °C (lit.)

SMILES string

CC(C)O[V](=O)(OC(C)C)OC(C)C

InChI

1S/3C3H7O.O.V/c3*1-3(2)4;;/h3*3H,1-2H3;;/q3*-1;;+3

InChI key

DSGGJXAUQHKOGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Vanadium(V) oxytriisopropoxide is a building block for a series of neutral oxovanadium complexes containing both ethoxide and acetylacetonate ligands the complexes have potential biological relevance as well as uses in catalysis. Vanadium (V) oxytriisopropoxide (VOTP) may be used directly as a V precursor to dope TiO2 to form a high performance anode for lithium ion batteries.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improving the electrochemical performance of anatase titanium dioxide by vanadium doping as an anode material for lithium-ion batteries.
Anh LT, et al.
Journal of Power Sources, 243, 891-898 (2013)
Feilong Jiang et al.
Inorganic chemistry, 37(21), 5439-5451 (2001-10-24)
The studies of an underexplored synthetic reagent, VO(2)(acac) (Hacac = acetylacetone) and semirational strategies for the formation of a complete series of simple vanadium(V) alkoxide clusters in alcohol-containing solvents. The neutral mono-, di-, tri-, and tetranuclear oxovanadium(V) complexes [V(2)O(2)(&mgr;-OCH(3))(2)(acac)(2)(OCH(3))(2)] (1)
Colm Glynn et al.
Nanoscale, 7(47), 20227-20237 (2015-11-18)
Devices composed of transparent materials, particularly those utilizing metal oxides, are of significant interest due to increased demand from industry for higher fidelity transparent thin film transistors, photovoltaics and a myriad of other optoelectronic devices and optics that require more

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service