Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

392626

Sigma-Aldrich

Diethyl (hydroxymethyl)phosphonate

technical grade

Synonym(s):

(Hydroxymethyl)phosphonic acid diethyl ester, Diethyl phosphonomethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2P(O)(OC2H5)2
CAS Number:
Molecular Weight:
168.13
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.444 (lit.)

bp

124-126 °C/3 mmHg (lit.)

density

1.14 g/mL at 25 °C (lit.)

functional group

hydroxyl
phosphonate

SMILES string

CCOP(=O)(CO)OCC

InChI

1S/C5H13O4P/c1-3-8-10(7,5-6)9-4-2/h6H,3-5H2,1-2H3

InChI key

RWIGWWBLTJLKMK-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • α And β-dialkoxyphosphoryl isothiocyanate used in antiproliferative studies
  • Dendritic polyglycerol anions for L-selectin inhibition
  • Phosphonates for anti-HIV-1 activity
  • Homo and copolymers of phosphonated norbornenes

Involved in studies of FBPase inhibition by phosphonic acid-containing thaizoles

Reactant for Mitsunobu reactions

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

>752.0 °F - closed cup

Flash Point(C)

> 400 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service