Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

414034

Sigma-Aldrich

3-Nitrophthalimide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4N2O4
CAS Number:
Molecular Weight:
192.13
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

97%

form

powder or crystals

mp

213-215 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[O-][N+](=O)c1cccc2C(=O)NC(=O)c12

InChI

1S/C8H4N2O4/c11-7-4-2-1-3-5(10(13)14)6(4)8(12)9-7/h1-3H,(H,9,11,12)

InChI key

BONIIQYTWOPUQI-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A C Begg et al.
The British journal of radiology, 58(691), 645-654 (1985-07-01)
Two compounds, nitroakridin 3582 (NA) and a 3-nitro-naphthalimide (DM113), have been tested as potential fluorescent markers for hypoxic cells. Cellular fluorescence in three cell lines (V79-379A, WHF1B, EMT6) was measured by flow cytometry and high-performance liquid chromatography (HPLC) after incubating
Jun Yang et al.
Biotechnology and applied biochemistry, 66(1), 33-42 (2018-09-20)
A nitroreductase-encoded gene from an efficient nitro-reducing bacterium Streptomyces mirabilis DUT001, named snr, was cloned and heterogeneously expressed in Escherichia coli. The purified Streptomyces nitroreductase SNR was a homodimer with an apparent subunit molecular weight of 24 kDa and preferred NADH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service