Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

413666

Sigma-Aldrich

Indium(III) iodide

anhydrous, powder, 99.998% trace metals basis

Synonym(s):

Indium triiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
InI3
CAS Number:
Molecular Weight:
495.53
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.998% trace metals basis
grade:
anhydrous
form:
powder

grade

anhydrous

Quality Level

Assay

99.998% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: indium

impurities

≤25.0 ppm Trace Metal Analysis

SMILES string

I[In](I)I

InChI

1S/3HI.In/h3*1H;/q;;;+3/p-3

InChI key

RMUKCGUDVKEQPL-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Used to prepare vinyl and methyl indates which undergo regioselective reactions with cinnamyl bromide.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hirashita, T., et al.
Tetrahedron Letters, 45, 3225-3225 (2004)

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service