Skip to Content
MilliporeSigma

394599

Magnesium iodide

98%

Synonym(s):

Magnesium diiodide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
MgI2
CAS Number:
Molecular Weight:
278.11
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12352302
EC Number:
233-825-1
MDL number:
Assay:
98%
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

98%

form

powder

reaction suitability

core: magnesium

mp

637 °C (dec.) (lit.)

density

4.43 g/mL at 25 °C (lit.)

SMILES string

I[Mg]I

InChI

1S/2HI.Mg/h2*1H;/q;;+2/p-2

InChI key

BLQJIBCZHWBKSL-UHFFFAOYSA-L

Application

When reacted with cyclic sulfates of vic-diols, produces the corresponding olefins in excellent yields.


Still not finding the right product?

Explore all of our products under Magnesium iodide


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library