Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41314

Sigma-Aldrich

2-Maleimidoethyl mesylate

technical, ≥90.0% (HPLC)

Synonym(s):

2-Maleimidoethyl methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9NO5S
Molecular Weight:
219.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

≥90.0% (HPLC)

reaction suitability

reagent type: cross-linking reagent

functional group

maleimide
mesylate

SMILES string

O=C(C=C1)N(CCOS(=O)(C)=O)C1=O

InChI

1S/C7H9NO5S/c1-14(11,12)13-5-4-8-6(9)2-3-7(8)10/h2-3H,4-5H2,1H3

InChI key

PMKKIDFHWBBGDA-UHFFFAOYSA-N

Application

Hetero-bifunctional linker specially for the quaternisation of pyridyl-nitrogen in order to introduce an amine or thiol reactive group. Synthetic building block for the preparation of fluorescent labels

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Litak, P.T.;Kauffman, J.M.
Journal of Heterocyclic Chemistry, 31, 457-457 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service