Skip to Content
MilliporeSigma

358657

3-Maleimidopropionic acid N-hydroxysuccinimide ester

99%

Synonym(s):

N-(3-Maleimidopropionyloxy)succinimide, N-Maleoyl-β-alanine N′-hydroxysuccinimide ester, N-Maleoyl-β-alanine N-hydroxysuccinimide ester, N-Succinimidyl 3-maleimidopropionate, BMPS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H10N2O6
CAS Number:
Molecular Weight:
266.21
UNSPSC Code:
12352106
NACRES:
NA.25
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1492578
Assay:
99%
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

99%

form

powder

reaction suitability

reagent type: linker

mp

168-170 °C (lit.)

functional group

NHS ester

storage temp.

2-8°C

SMILES string

O=C1CCC(N1OC(CCN2C(C=CC2=O)=O)=O)=O

InChI

1S/C11H10N2O6/c14-7-1-2-8(15)12(7)6-5-11(18)19-13-9(16)3-4-10(13)17/h1-2H,3-6H2

InChI key

JKHVDAUOODACDU-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library