Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

407518

Sigma-Aldrich

Julolidine hydrobromide

97%

Synonym(s):

2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizine hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15N · HBr
CAS Number:
Molecular Weight:
254.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

239-242 °C (lit.)

SMILES string

Br.C1CN2CCCc3cccc(C1)c23

InChI

1S/C12H15N.BrH/c1-4-10-6-2-8-13-9-3-7-11(5-1)12(10)13;/h1,4-5H,2-3,6-9H2;1H

InChI key

KHWBRFVBPGPEOJ-UHFFFAOYSA-N

General description

Julolidines are effective auxofluors that are employed in laser dyes and biochemical stains. They are known to be one of the strongest electron-releasing groups due to electronic and steric factors.

Application

Julolidine hydrobromide may be used in the synthesis of [4-(2,3,6,7-tetrahydro-1H,5H-pyrido[3,2,1-ij]quinolin-9-ylazo)-phenyl]-methanol azodye.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, optical characterization and crystal and molecular X-ray structure of a phenylazojulolidine derivative.
Barbero N, et al.
Dyes and Pigments, 92(3), 1177-1183 (2012)
Synthesis of julolidine derivatives.
Kauffman JM, et al.
Organic preparations and procedures international, 33(6), 603-613 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service