Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C16H14N2O3
CAS Number:
Molecular Weight:
282.29
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
243-569-2
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
solubility
DMSO: soluble ≥1 mg/mL, methanol: soluble ≥1 mg/mL
storage temp.
2-8°C
SMILES string
OC(COC1=NN(CC2=CC=CC=C2)C3=CC=CC=C31)=O
InChI
1S/C16H14N2O3/c19-15(20)11-21-16-13-8-4-5-9-14(13)18(17-16)10-12-6-2-1-3-7-12/h1-9H,10-11H2,(H,19,20)
InChI key
BYFMCKSPFYVMOU-UHFFFAOYSA-N
Gene Information
human ... XDH(7498)
Biochem/physiol Actions
Bendazac (or bendazolic acid) is a non-steroidal anti-inflammatory drug used for joint and muscular pain. Bendazac is an anti-denaturant drug for cataract and other condensation diseases. In addition, Bendazac has been used in studies related to potential treatment and pevention of ischemia and reperfusion syndromes, and was found to inhibit only reversible and irreversible xanthine oxidase.
Bendazac is a non-steroidal anti-inflammatory drug used for joint and muscular pain.
Storage Class
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.