Skip to Content
MilliporeSigma
All Photos(2)

Documents

400688

Sigma-Aldrich

Sodium tellurite

−100 mesh, 99%

Synonym(s):

Disodium trioxotellurate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2TeO3
CAS Number:
Molecular Weight:
221.58
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

form

powder

particle size

−100 mesh

SMILES string

[Na+].[Na+].[O-][Te]([O-])=O

InChI

1S/2Na.H2O3Te/c;;1-4(2)3/h;;(H2,1,2,3)/q2*+1;/p-2

InChI key

VOADVZVYWFSHSM-UHFFFAOYSA-L

General description

Sodium terllurite is an alkali tellurite that is majorly used in the formation of glasses due to its unique properties which include good transmittance, refractive index, and high chemical resistance. It can be used as a tellurite source for the preparation of quantum dots.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marketa Ryvolova et al.
Electrophoresis, 32(13), 1619-1622 (2011-05-25)
In this study, biotin-conjugated glutathione was synthesized using peptide bonding of the biotin carboxy group and amino group of the γ-glutamic acid to prepare an alternative coating for CdTe quantum dots (QDs). This type of coating combines the functionality of
Jingwen Gao et al.
Talanta, 199, 178-183 (2019-04-07)
In this work, an innovative aptasensor based on electrochemiluminescence resonance energy transfer (ECL-RET) from CdTe quantum dots (QDs) to a cyanine dye (Cy5) fluorophore for the determination of Ochratoxin A (OTA) was fabricated. A strong cathodic ECL emission was obtained
Pavlina Sobrova et al.
International journal of molecular sciences, 14(7), 13497-13510 (2013-06-29)
Electrochemical detection of quantum dots (QDs) has already been used in numerous applications. However, QDs have not been well characterized using voltammetry, with respect to their characterization and quantification. Therefore, the main aim was to characterize CdTe QDs using cyclic
Structural properties and crystallization of sodium tellurite glasses
Santic A, et al.
Croatica Chemica Acta. Arhiv Za Kemiju, 81(4), 559-567 (2008)
Animesh Jha et al.
Optics express, 16(18), 13526-13533 (2008-09-06)
In this paper we report the effect of heat treatment of 10 mol% Na(2)O-89.5 mol% TeO(2) glass for studying the fluorescence and line broadening behavior of Er-ions in the glass-ceramic matrix. Controlled crystal growth measurements using the X-ray line broadening

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service