Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

393916

Sigma-Aldrich

2-Ethylphenylhydrazine hydrochloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C6H4NHNH2·HCl
CAS Number:
Molecular Weight:
172.66
Beilstein:
3697547
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

178 °C (dec.) (lit.)

solubility

water: soluble

functional group

hydrazine

SMILES string

Cl[H].CCc1ccccc1NN

InChI

1S/C8H12N2.ClH/c1-2-7-5-3-4-6-8(7)10-9;/h3-6,10H,2,9H2,1H3;1H

InChI key

HBHPTOKYVGZBAJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Ethylphenylhydrazine hydrochloride is a hydrazine derivative.

Packaging

2-Ethylphenylhydrazine hydrochloride may be used in the synthesis of 7-ethyltryptophol, an intermediate for the anti-inflammmatory drug etodolac.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An improved and scalable process for the synthesis of a key intermediate for Etodolac, a non-steroidal anti-inflammatory drug
Sekharayya MC, et al.
Indian J. Chem. B, 51(12), 1763-1766 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service