Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T40606

Sigma-Aldrich

p-Tolylhydrazine hydrochloride

98%

Synonym(s):

4-Methylphenylhydrazine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4NHNH2 · HCl
CAS Number:
Molecular Weight:
158.63
Beilstein:
3563996
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

>200 °C (dec.) (lit.)

SMILES string

Cl.Cc1ccc(NN)cc1

InChI

1S/C7H10N2.ClH/c1-6-2-4-7(9-8)5-3-6;/h2-5,9H,8H2,1H3;1H

InChI key

HMHWNJGOHUYVMD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Toth et al.
Journal of toxicology and environmental health, 8(1-2), 1-9 (1981-07-01)
4-Methylphenylhydrazine hydrochloride (4-MPH) was administered to randomly bred Swiss mice as 26 weekly sc injections of 140 microgram per gram of body weight and N'-acetyl-4-(hydroxymethyl)phenylhydrazine (AMPH) as 26 weekly sc injections of 500 microgram/g. As a solvent control, physiological saline
Megumi Hamano Nagaokaa et al.
Chemical & pharmaceutical bulletin, 54(6), 922-924 (2006-06-07)
Agaricus blazei MURRILL mushroom products are sold as so-called health foods in Japan. However, a part of Agaricus is known to contain hydrazines. A sensitive and specific method for analyzing a genotoxic hydrazine, agaritine, and its derivatives was developed to
Qiang Tang et al.
Journal of the American Chemical Society, 130(18), 5840-5841 (2008-04-15)
An unexpected reaction of N,N-disubstituted hydrazine with naphthol and its analogues under simply thermal conditions has been disclosed. 2-Naphthol reacted with various N,N-disubstituted hydrazines under argon to afford 1-amino-2-naphthol and the corresponding secondary amines in excellent yields. Ortho amination of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service