Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

393398

Sigma-Aldrich

Tetramethyl-d12 orthosilicate

99 atom % D

Synonym(s):

Tetramethoxy-d12-silane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(OCD3)4Si
CAS Number:
Molecular Weight:
164.30
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.23

isotopic purity

99 atom % D

Quality Level

refractive index

n20/D 1.366 (lit.)

density

1.114 g/mL at 25 °C (lit.)

mass shift

M+12

SMILES string

[2H]C([2H])([2H])O[Si](OC([2H])([2H])[2H])(OC([2H])([2H])[2H])OC([2H])([2H])[2H]

InChI

1S/C4H12O4Si/c1-5-9(6-2,7-3)8-4/h1-4H3/i1D3,2D3,3D3,4D3

InChI key

LFQCEHFDDXELDD-MGKWXGLJSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Silica's versatility spans various industries, including biomedical applications.

Silica's versatility spans various industries, including biomedical applications.

Silica's versatility spans various industries, including biomedical applications.

Silica's versatility spans various industries, including biomedical applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service