Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

392456

Sigma-Aldrich

2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine ruthenium(II) carbonyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H44N4ORu
CAS Number:
Molecular Weight:
661.84
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

reaction suitability

reagent type: catalyst
core: ruthenium

λmax

391 nm

SMILES string

[C-]#[O+].CCc1c(CC)c2cc3c(CC)c(CC)c4cc5nc(cc6c(CC)c(CC)c(cc1n2)n6[Ru]n34)c(CC)c5CC

InChI

1S/C36H44N4.CO.Ru/c1-9-21-22(10-2)30-18-32-25(13-5)26(14-6)34(39-32)20-36-28(16-8)27(15-7)35(40-36)19-33-24(12-4)23(11-3)31(38-33)17-29(21)37-30;1-2;/h17-20H,9-16H2,1-8H3;;/q-2;;+2/b29-17-,30-18-,31-17-,32-18-,33-19-,34-20-,35-19-,36-20-;;

InChI key

CRTFSNIGJRIMPF-YAJYDHHFSA-N

Application

2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine ruthenium(II) carbonyl (RuOEP )can form blended polymeric films with poly (3-hexythiopene-2,5-diyl)( P3HT).

Other Notes

Contains solvent of crystallization

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inorgorganica Chimica Acta, 85, 209-209 (1984)
Aldrichimica Acta, 19, 81-81 (1986)
Am. Chem. Soc. Symp. Ser. No. 152, 243-243 (1981)
2D-IR Studies of Annealing-Induced Changes to Structural Dynamics in Organic Semiconductor Thin Films.
Eigner AA, et al.
The Journal of Physical Chemistry C, 114(28), 12308-12315 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service