Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

298980

Sigma-Aldrich

o-Tolylmagnesium bromide solution

2.0 M in diethyl ether

Synonym(s):

2-Methylphenylmagnesium bromide, 2-Tolylmagnesium bromide, o-Methylphenylmagnesium bromide, Bromo(2-methylphenyl)magnesium, Bromo-o-tolylmagnesium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4MgBr
CAS Number:
Molecular Weight:
195.34
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Grignard Reaction

concentration

2.0 M in diethyl ether

density

1.013 g/mL at 25 °C

SMILES string

Cc1ccccc1[Mg]Br

InChI

1S/C7H7.BrH.Mg/c1-7-5-3-2-4-6-7;;/h2-5H,1H3;1H;/q;;+1/p-1

InChI key

DVXDIGKNJYSMFM-UHFFFAOYSA-M

Application

o-Tolylmagnesium bromide solution can be used to prepare:
  • An iridium(I) mesityl complex named Ir(o-tolyl)(CO)(dppe).
  • 4-(2-Methylphenyl)-4-oxobutanoic acid, a key intermediate for the synthesis of M1 muscarinic selective agonist AC-42 derivatives.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-40.0 °F

Flash Point(C)

-40 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chihiro Tsukano et al.
Chemical & pharmaceutical bulletin, 65(12), 1167-1174 (2017-12-05)
Tetrahydrobiphenylene consists of cyclobutene fused with benzene and cyclohexene rings. In this paper, a direct method for synthesizing tetrahydrobiphenylenes based on a palladium (Pd)(0)-catalyzed C(sp
Synthesis and Reactivity of the Iridium (I) Mesityl Complex Ir (CO)(mes)(dppe). Oxidative Addition and Ligand Activation Reactions
Cleary BP and Eisenberg R
Journal of the American Chemical Society, 117(12), 3510-3521 (1995)
Fluorescent derivatives of AC-42 to probe bitopic orthosteric/allosteric binding mechanisms on muscarinic M1 receptors
Daval SB, et al.
Journal of Medicinal Chemistry, 55(5), 2125-2143 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service