391824
Zirconyl nitrate solution
35 wt. % in dilute nitric acid, ≥99% trace metals basis
Synonym(s):
Bis(nitrato)oxozirconium, Zirconium oxide nitrate
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
ZrO(NO3)2
CAS Number:
Molecular Weight:
231.23
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
liquid
Recommended Products
grade
for analytical purposes
Assay
≥99% trace metals basis
form
liquid
concentration
35 wt. % in dilute nitric acid
density
1.45 g/mL at 25 °C
application(s)
battery manufacturing
SMILES string
O=[Zr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
InChI
1S/2NO3.O.Zr/c2*2-1(3)4;;/q2*-1;;+2
InChI key
JWBWFTANBHYHBC-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 2 - Skin Corr. 1B
Supplementary Hazards
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Pravin S Shinde et al.
Scientific reports, 6, 32436-32436 (2016-09-01)
Augmenting the donor density and nanostructure engineering are the crucial points to improve solar water oxidation performance of hematite (α-Fe2O3). This work addresses the sluggish water oxidation reaction associated with hematite photoanode by tweaking its internal porosity. The porous hematite
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service