Skip to Content
MilliporeSigma
All Photos(1)

Documents

389188

Sigma-Aldrich

Glyceryl tribenzoate

95%

Synonym(s):

Tribenzoin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CO2CH(CH2O2CC6H5)2
CAS Number:
Molecular Weight:
404.41
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

mp

68-72 °C (lit.)

SMILES string

O=C(OCC(COC(=O)c1ccccc1)OC(=O)c2ccccc2)c3ccccc3

InChI

1S/C24H20O6/c25-22(18-10-4-1-5-11-18)28-16-21(30-24(27)20-14-8-3-9-15-20)17-29-23(26)19-12-6-2-7-13-19/h1-15,21H,16-17H2

InChI key

HIZCTWCPHWUPFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Plasticizer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Li et al.
PloS one, 16(2), e0247236-e0247236 (2021-02-20)
Berberine is a promising botanical pesticide against fungal plant pathogens. However, whether berberine inhibits the invasion of fungal pathogen across plant surface remains unclear. Here we demonstrated that the enzyme activities of purified cutinase from fungal pathogen Colletotrichum capsici were
Ying Dong et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 957, 30-35 (2014-03-25)
Glycerol has the latent capacity to act as a plasma volume expander and disguise blood doping practices. Therefore, it has been prohibited in sports as a masking agent by the World Anti-Doping Agency (WADA) since January 2010 and a urinary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service