Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

387738

Sigma-Aldrich

4-Hydroxyphenylacetamide

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH2CONH2
CAS Number:
Molecular Weight:
151.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

mp

176-178 °C (lit.)

functional group

amide

SMILES string

NC(=O)Cc1ccc(O)cc1

InChI

1S/C8H9NO2/c9-8(11)5-6-1-3-7(10)4-2-6/h1-4,10H,5H2,(H2,9,11)

InChI key

YBPAYPRLUDCSEY-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Owino et al.
Journal of chromatographic science, 29(10), 450-456 (1991-10-01)
The diode array multichannel detector is used to acquire spectral information at specified intervals in the elution profile of atenolol and its related impurities for post-chromatographic data analysis. The applicability of a number of peak homogeneity testing methods, including spectral
Gerd Sigmund et al.
Drug testing and analysis, 7(11-12), 1057-1062 (2015-10-21)
2-Phenylethanamine (phenethylamine, PEA) represents the core structure of numerous drugs with stimulant-like properties and is explicitly featured as so-called specified substance on the World Anti-Doping Agency (WADA) Prohibited List. Due to its natural occurrence in humans as well as its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service