Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

378488

Sigma-Aldrich

Poly(dimethylsiloxane-co-methylphenylsiloxane)

viscosity 125 cSt 

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Si(CH3)2O]x[Si(CH3)(C6H5)O]y
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

viscous liquid

Quality Level

refractive index

n20/D 1.497

viscosity

125 cSt

density

1.066 g/mL at 25 °C

SMILES string

C[Si](C)=O.C[Si](=O)c1ccccc1

InChI

1S/C7H8OSi.C2H6OSi/c1-9(8)7-5-3-2-4-6-7;1-4(2)3/h2-6H,1H3;1-2H3

InChI key

XWFOTLCDICUQTI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

437.0 °F - closed cup

Flash Point(C)

225.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service