378445
Poly(dimethylsiloxane-co-methylphenylsiloxane)
viscosity 50 cSt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
viscous liquid
Quality Level
refractive index
n20/D 1.423
viscosity
50 cSt
density
0.982 g/mL at 25 °C
SMILES string
C[Si](C)=O.C[Si](=O)c1ccccc1
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
>214.0 °F - closed cup
Flash Point(C)
> 101.1 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service