Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

377031

Sigma-Aldrich

Butyl glycidyl ether

95%

Synonym(s):

(Butoxymethyl)oxirane, Araldite® RD-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14O2
CAS Number:
Molecular Weight:
130.18
Beilstein:
103668
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

liquid

refractive index

n20/D 1.418 (lit.)

bp

164-166 °C (lit.)

density

0.91 g/mL at 25 °C (lit.)

SMILES string

CCCCOCC1CO1

InChI

1S/C7H14O2/c1-2-3-4-8-5-7-6-9-7/h7H,2-6H2,1H3

InChI key

YSUQLAYJZDEMOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butyl glycidyl ether is an epoxy monomer that is mainly used as an aliphatic reactive diluent in the modification of epoxy systems. Its structure consists of a soft segment and one epoxy segment which can be incorporated into the epoxy resins for reducing the viscosity. It facilitates and enhances the mechanical properties by improving the cryogenic strength, ductility and impact resistance of the polymers.

Legal Information

Araldite is a registered trademark of Huntsman Advanced Materials Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Repr. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and characterization of layered silicate/glass fiber/epoxy hybrid nanocomposites via vacuum-assisted resin transfer molding (VARTM)
Lin L, et al.
Composites Science and Technology, 66(13), 2116-2125 (2006)
Mechanical property characterization of a polymeric nanocomposite reinforced by graphitic nanofibers with reactive linkers
Xu LR, et al.
Journal of Composite Materials, 38(18), 1563-1582 (2004)
T H Connor et al.
Environmental mutagenesis, 2(4), 521-530 (1980-01-01)
The epoxide diluent, n-butylglycidyl ether (n-BGE), was tested in vitro for mutagenicity using Salmonella typhimurium and for transforming ability in mouse embryo cells (BAlb/3T3-A31-1-13). The n-BGE produced base-pair type mutations in S typhimurium strains TA1535 and TA100. However, it failed
L-J Chen et al.
Drug metabolism and disposition: the biological fate of chemicals, 35(12), 2218-2224 (2007-09-19)
The disposition of [(14)C]-labeled n-butyl glycidyl ether (BGE, 3-butoxy-1,2-epoxypropane) was studied in rats and mice. The majority of a single p.o. dose (2-200 mg/kg) was excreted in urine (rats, 84-92%; mice, 64-73%) within 24 h. The rest of the dose
Mohammad F Bostanudin et al.
Polymers, 11(5) (2019-05-06)
Pectin is a polysaccharide with very good gel forming properties that traditionally has found important applications in foods and pharmaceutical industries. Although less studied, chemical modifications of pectin leading to a decrease in its hydrophilicity can be useful for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service