Skip to Content
MilliporeSigma
All Photos(1)

Documents

251712

Sigma-Aldrich

tert-Butyl glycidyl ether

99%

Synonym(s):

(tert-Butoxymethyl)oxirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14O2
CAS Number:
Molecular Weight:
130.18
Beilstein:
103483
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.417 (lit.)

density

0.917 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)OCC1CO1

InChI

1S/C7H14O2/c1-7(2,3)9-5-6-4-8-6/h6H,4-5H2,1-3H3

InChI key

SFJRUJUEMVAZLM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Sens. 1

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Kotik et al.
Biochimica et biophysica acta, 1760(2), 245-252 (2005-12-14)
Purification of a novel enantioselective epoxide hydrolase from Aspergillus niger M200 has been achieved using ammonium sulphate precipitation, ionic exchange, hydrophobic interaction, and size-exclusion chromatography, in conjunction with two additional chromatographic steps employing hydroxylapatite, and Mimetic Green. The enzyme was
A F Frost et al.
Mutation research, 102(2), 193-200 (1982-09-01)
Unscheduled DNA synthesis (UDS) was examined in normal human peripheral blood lymphocytes (HPBL) exposed to n- and tert.-butyl glycidyl ethers (BGE). n-BGE, which is currently used as a major reactive diluent for industrial epoxy resin systems, has been implicated as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service