Skip to Content
MilliporeSigma
All Photos(1)

Documents

368857

Sigma-Aldrich

p-Tolyl ether

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3C6H4)2O
CAS Number:
Molecular Weight:
198.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

bp

285 °C (lit.)

mp

47-49 °C (lit.)

SMILES string

Cc1ccc(Oc2ccc(C)cc2)cc1

InChI

1S/C14H14O/c1-11-3-7-13(8-4-11)15-14-9-5-12(2)6-10-14/h3-10H,1-2H3

InChI key

YWYHGNUFMPSTTR-UHFFFAOYSA-N

Related Categories

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

XII.-Substituted diaryl ethers. Part I. Di-p-tolyl ether.
BoydaBarrett HS.
Journal of the Chemical Society, 67-73 (1927)
Solid-State 13C NMR, X-ray, and Quantum Mechanical Studies of the Carbon Chemical Shifts Tensors of p-Tolyl Ether.
Facelli JC, et al.
The Journal of Physical Chemistry, 98(47), 12186-12190 (1994)
Anders Lennartson et al.
Chirality, 27(7), 425-429 (2015-06-03)
We studied the spontaneous formation of chiral crystals of four diaryl ethers, 3-phenoxybenzaldehyde, 1; 1,3-dimethyl-2-phenoxybenzene, 2; di(4-aminophenyl) ether, 3; and di(p-tolyl) ether, 4. Compounds 1, 3, and 4 form conformationally chiral molecules in the solid state, while the chirality of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service