Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

367818

Sigma-Aldrich

1,1′-Sulfonyldiimidazole

98%

Synonym(s):

1,1′-Sulfonylbis[1H-imidazole], 1-Imidazol-1-ylsulfonylimidazole, N,N′-Sulfuryldiimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N4O2S
CAS Number:
Molecular Weight:
198.20
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

135-137 °C (lit.)

SMILES string

O=S(=O)(n1ccnc1)n2ccnc2

InChI

1S/C6H6N4O2S/c11-13(12,9-3-1-7-5-9)10-4-2-8-6-10/h1-6H

InChI key

ZLKNPIVTWNMMMH-UHFFFAOYSA-N

General description

The mol­ecule of 1,1′-sulfonyldiimidazole, is reported to possess a pseudo-twofold rotation axis of symmetry passing through the S atom. It undergoes one pot Suzuki Coupling reaction to afford 1-phenylnaphthalene.

Application

1,1′-Sulfonyldiimidazole may be used in the preparation of stable 1:1 complexes of benziodoxole. It may be used in the synthesis of 2,3′-anhydro-2′-deoxyribonucleosides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphane-free Suzuki-Miyaura coupling of aryl imidazolesulfonates with arylboronic acids and potassium aryltrifluoroborates under aqueous conditions.
Civicos JF, et al.
Chemistry Letters (Jpn), 40(9), 907-909 (2011)
A facile one-pot synthesis of 2, 3'-anhydro-2'-deoxyuridines via 3'-O-imidazolylsulfonates.
No Z, et al.
Synthetic Communications, 30(21), 3873-3882 (2000)
1, 1'-Sulfonyldiimidazole.
Patel UH, et al.
Acta Crystallographica Section E, Structure Reports Online, 63(8), o3598-o3599 (2007)
Complexes of hypervalent iodine compounds with nitrogen ligands.
Zhdankin VV, et al.
Tetrahedron, 43(33), 5735-5737 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service