Skip to Content
MilliporeSigma
All Photos(1)

Documents

338931

Sigma-Aldrich

3,4,5-Trimethoxybenzylamine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3C6H2CH2NH2
CAS Number:
Molecular Weight:
197.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.548 (lit.)

bp

121 °C/0.05 mmHg (lit.)

density

1.155 g/mL at 25 °C (lit.)

SMILES string

COc1cc(CN)cc(OC)c1OC

InChI

1S/C10H15NO3/c1-12-8-4-7(6-11)5-9(13-2)10(8)14-3/h4-5H,6,11H2,1-3H3

InChI key

YUPUSBMJCFBHAP-UHFFFAOYSA-N

Application

3,4,5-Trimethoxybenzylamine was used in the synthesis of benzyl 3-deoxy-3-(3,4,5-trimethoxybenzylamino)-β-L−xylopyranoside.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benzyl 3-deoxy-3-(3, 4, 5-trimethoxybenzylamino)-?-L-xylopyranoside.
Benzyl 3-deoxy-3-(3, 4, 5-trimethoxybenzylamino)-?-L-xylopyranoside.
Molbank, 2013(1), M793-M793 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service