Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

337439

Sigma-Aldrich

Tri-O-benzyl-D-glucal

97%

Synonym(s):

(3R,4S,5R)-3,4,6-Tri-O-benzyl-D-glucal, 1,5-Anhydro-2-deoxy-3,4,6-tris-O-(phenylmethyl)-D-arabino-hex-1-enitol, 3,4,6-Tri-O-benzyl-D-glucal, Tri-OBn-glucal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H28O4
CAS Number:
Molecular Weight:
416.51
Beilstein:
1691968
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]25/D −2.7°, c = 5 in chloroform

mp

57-58 °C (lit.)

storage temp.

−20°C

SMILES string

C(OCc1ccccc1)[C@H]2OC=C[C@@H](OCc3ccccc3)[C@@H]2OCc4ccccc4

InChI

1S/C27H28O4/c1-4-10-22(11-5-1)18-28-21-26-27(31-20-24-14-8-3-9-15-24)25(16-17-29-26)30-19-23-12-6-2-7-13-23/h1-17,25-27H,18-21H2/t25-,26-,27+/m1/s1

InChI key

MXYLLYBWXIUMIT-PFBJBMPXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Important building block for both solution- and solid-phase synthesis of oligosaccharides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Seeberger, P.H., et al.
Aldrichimica Acta, 30, 75-75 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service